Learn More
Bio X Cell Monoclonal Antibody, Human CD71, OKT-9, 1 MG
Supplier: Bio X Cell BE0023R001MG
The OKT-9 monoclonal antibody reacts with human CD71, also known as Transferrin receptor protein 1 (TfR1). CD71 is a 95 kDa type II transmembrane glycoprotein involved in iron uptake and is a potential new target in human leukemia and lymphoma.
- Clone: OKT-9
- Reactivities: Human
- Isotype: Mouse IgG1, κ
- Unconjugated (conjugation services available)
- Immunogen: Human acute lymphoblastic leukemia cells
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Purification method: Protein G (animal-free facility)
- Reported applications: Immunohistochemistry (frozen), immunofluorescence, flow cytometry
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.