Learn More
Bio X Cell anti-rat/mouse CD71, OX-26 IgG2a, Unconjugated, 1 MG
Supplier: Bio X Cell BE0331R001MG
This InVivoMAb anti-rat/mouse CD71 (TfR1) antibody, clone OX-26, Mouse IgG2a, reacts with rat CD71 and cross-reacts with mouse CD71. It is frequently utilized for targeted drug delivery to the brain.
- Reacts with rat CD71 (TfR1) and cross-reacts with mouse CD71
- Isotype: IgG2a, κ
- Applications include targeted drug delivery to the brain, immunohistochemistry (frozen), and flow cytometry
- Purified from cell culture supernatant in an animal-free facility using Protein A
- Formulation: PBS, pH 7.0, with no stabilizers or preservatives
- Store at stock concentration at 4°C; do not freeze
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg
- Sterility: 0.2 μm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.