Learn More
Bio X Cell InVivoMAb anti-human CEACAM7, Clone P3-7B, Mouse IgG1, Unconjugated, 1 MG
Supplier: Bio X Cell BE0363R001MG
This monoclonal antibody (clone P3-7B) reacts with human carcinoembryonic antigen-related cell adhesion molecule 7 (CEACAM7). CEACAM7 is a highly glycosylated membrane protein belonging to the immunoglobulin superfamily, known for its expression in columnar epithelial cells of the large intestine. This antibody is frequently employed in identifying cells expressing this glycoprotein in tissue samples and cell cultures.
- Reacts with human CEACAM7
- Suitable for western blot
- Suitable for ELISA
- Suitable for immunoprecipitation
- Suitable for immunohistochemistry
- Suitable for flow cytometry
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level of 1EU/mg
- Purity of 95% by SDS-PAGE
- Sterilized by 0.2 m filtration
- Purified by protein G
- Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.