Learn More
Bio X Cell CEACAM7 Antibody, P3-7B, Mouse IgG1, 50 MG
Supplier: Bio X Cell BE0363R050MG
The P3-7B monoclonal antibody targets human carcinoembryonic antigen-related cell adhesion molecule 7 (CEACAM7). CEACAMs are glycosylated membrane proteins. Antibodies against CEACAMs are commonly used to identify cells expressing the glycoprotein in tissue and cell culture samples.
- Isotype: Mouse IgG1, κ
- Immunogen: Human CEACAM7-Fc fusion protein
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Production: Purified in an animal-free facility
- Purification method: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.