Learn More
Bio X Cell Anti-mouse CD172a P84 Antibody, 5 MG
Supplier: Bio X Cell BE0322R005MG
P84 monoclonal antibody targets Signal-Regulatory Protein α (SIRPα, CD172a), a glycoprotein on immune cells and neurons. It regulates T cell activation, neutrophil migration, and phagocytosis by binding CD47, demonstrating neutralizing activity in vivo and in vitro.
- Clone: P84
- Reactivities: mouse
- Isotype: rat IgG1, κ
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in animal-free facility
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
- Immunogen: mouse brain membrane protein
- Applications: in vivo and in vitro SIRPα blocking, Western blot, Immunoprecipitation, Flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.