Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Monoclonal Antibody, anti-mouse NK1.1, PK136, 1 MG

Catalog No. 501528267
Encompass_Preferred

The PK136 monoclonal antibody reacts with mouse NK1.1, also known as CD161b/CD161c, KLRB1, NKR-P1A, and Ly-55. NK1.1 is a type II integral membrane glycoprotein with a C-type lectin domain, encoded by the Klrb1c/NKR-P1C gene.

  • Reported applications: In vivo NK cell depletion, flow cytometry
  • Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein A
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/µg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 µm filtration
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Catalog No. 50-152-8267 Supplier Bio X Cell Supplier No. BE0036R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PK136 monoclonal antibody reacts with mouse NK1.1, also known as CD161b/CD161c, KLRB1, NKR-P1A, and Ly-55. NK1.1 is a type II integral membrane glycoprotein with a C-type lectin domain, encoded by the Klrb1c/NKR-P1C gene.

  • Reported applications: In vivo NK cell depletion, flow cytometry
  • Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein A
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/µg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 µm filtration
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.