Learn More
Bio X Cell Monoclonal Antibody, Clone 3H3, Mouse 4-1BB (CD137), Rat IgG2a, 100 MG
Supplier: Bio X Cell BP0239R100MG
The Bio X Cell InVivoPlus anti-mouse 4-1BB (CD137) monoclonal antibody, Clone 3H3, reacts with mouse 4-1BB (CD137), a transmembrane protein. This agonistic antibody stimulates 4-1BB signaling in vivo, providing costimulatory signals to CD4 and CD8 T cells. It is formulated in PBS, pH 7.0, with a purity of 95% and endotoxin level of 0.5EU/mg.
- Isotype: rat IgG2a
- Unconjugated format
- Immunogen: mouse CD137 human Fc fusion protein
- Reported for in vivo and in vitro 4-1BB stimulation
- Sterilized by 0.2 m filtration
- Produced in animal-free facility
- Purified by protein G
- Aggregation less than 5%
- Molecular weight: 150 kDa
- Store at 4C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.