Learn More
Bio X Cell Armenian Hamster IgG, 95 Percent, 1 MG
Supplier: Bio X Cell BE0091R001MG
This polyclonal Armenian hamster IgG is purified from Armenian hamster serum and serves as a non-reactive control IgG for Armenian hamster antibodies in various in vivo and in vitro applications.
- Isotype: Armenian hamster IgG
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1EU/mg, determined by LAL assay
- Sterility: 0.2 μm filtration
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.