Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Polyclonal Human IgG, Unconjugated, 1 MG

Catalog No. 501528312
Encompass_Preferred

This polyclonal human IgG is purified from human serum and is suitable as a non-reactive control IgG for human IgG antibodies in most in vivo and in vitro applications.

  • Isotype: Human IgG
  • Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Purification: Protein A
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Catalog No. 50-152-8312 Supplier Bio X Cell Supplier No. BE0092R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This polyclonal human IgG is purified from human serum and is suitable as a non-reactive control IgG for human IgG antibodies in most in vivo and in vitro applications.

  • Isotype: Human IgG
  • Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Purification: Protein A
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.