Learn More
Bio X Cell InVivoMAb Polyclonal Rat IgG, Unconjugated, 1 MG
Supplier: Bio X Cell BE0094R001MG
This polyclonal rat IgG is purified from rat serum and serves as a non-reactive control for polyclonal rat IgG antibodies in various in vivo and in vitro applications.
- Isotype: Rat IgG
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Purification method: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.