Learn More
Bio X Cell InVivoMAb, Polyclonal Syrian Hamster IgG, 1 MG
Supplier: Bio X Cell BE0087R001MG
This polyclonal Syrian hamster IgG is purified from Syrian hamster serum, serving as a non-reactive control for Syrian hamster antibodies in most in vivo and in vitro applications.
- Isotype: Syrian hamster IgG
- Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
- Unconjugated
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification method: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.