Learn More
Bio X Cell Monoclonal Antibody, Clone PS/2, Mouse/Human, Rat IgG2b, 2 1 MG
Supplier: Bio X Cell BE0071R001MG
The PS/2 monoclonal antibody targets human and mouse VLA-4 chain (CD49d and integrin alpha 4), a 150 kDa glycoprotein of the integrin family. It is expressed by various cell types, including T and B lymphocytes, monocytes, eosinophils, basophils, mast cells, thymocytes, NK cells, and dendritic cells. This antibody is effective for in vivo and in vitro VLA-4 neutralization.
- Clone PS/2
- Reacts with mouse and human VLA-4
- Rat IgG2b, isotype
- Suitable for in vivo and in vitro VLA-4 neutralization
- Applicable for flow cytometry
- Purity of 95% ensures reliable experimental results
- Low endotoxin level of 1EU/mg
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Sterilized by 0.2 m filtration
- Purified using protein G from animal-free cell culture supernatant
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.