Learn More
Bio X Cell anti-mouse IL-12, R1-5D9, Rat IgG2a, 1 MG
Supplier: Bio X Cell BE0052R001MG
The R1-5D9 antibody reacts with mouse IL-12, a heterodimeric cytokine composed of subunits IL-12α p35 and IL-12β p40. This cytokine is secreted by activated monocytes, macrophages, and dendritic cells, playing roles in T lymphocyte differentiation, IFNγ production, and NK cell cytotoxicity. It is intended for in vivo IL-12 neutralization.
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 µm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.