Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-mouse CD71, R17 217.1.3/TIB-219, 1 MG

Catalog No. 501528375
Encompass_Preferred

The R17 217.1.3/TIB-219 monoclonal antibody reacts with mouse CD71 (TfR1), a 170-180 kDa homodimeric transmembrane glycoprotein essential for cellular proliferation and iron import.

  • Clone: R17 217.1.3/TIB-219
  • Isotype: Rat IgG2a, κ
  • Host species: Rat
  • Target species: Mouse CD71 (TfR1)
  • Applications: In vivo depletion of CD71+ cells, antibody-oligonucleotide conjugate (AOC) synthesis
  • Immunogen: Mouse erythroleukemia cell line 745.6
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Antibody solution should be stored at 4°C; do not freeze.

Catalog No. 50-152-8375 Supplier Bio X Cell Supplier No. BE0175R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R17 217.1.3/TIB-219 monoclonal antibody reacts with mouse CD71 (TfR1), a 170-180 kDa homodimeric transmembrane glycoprotein essential for cellular proliferation and iron import.

  • Clone: R17 217.1.3/TIB-219
  • Isotype: Rat IgG2a, κ
  • Host species: Rat
  • Target species: Mouse CD71 (TfR1)
  • Applications: In vivo depletion of CD71+ cells, antibody-oligonucleotide conjugate (AOC) synthesis
  • Immunogen: Mouse erythroleukemia cell line 745.6
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Antibody solution should be stored at 4°C; do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.