Learn More
Bio X Cell Antibody, CD11a R7-1, Mouse IgG1, 1 MG
Supplier: Bio X Cell BE0192R001MG
The R7-1 monoclonal antibody targets human LFA-1α (lymphocyte function-associated antigen 1 alpha), also known as integrin alpha L chain and CD11a. This antibody specifically identifies an epitope within the I-domain of LFA-1α. LFA-1 is crucial for leukocyte intercellular adhesion and participates in lymphocyte costimulatory signaling. The R7-1 antibody acts as a neutralizing antibody.
- Reactivities: Human
- Isotype: Mouse IgG1
- Conjugation: Unconjugated
- Reported applications: Functional assays, flow cytometry
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.