Learn More
Bio X Cell anti-mouse CD27, RM27-3E5, IgG2a, Unconjugated, 1 MG
Supplier: Bio X Cell BE0348R001MG
The RM27-3E5 monoclonal antibody reacts with mouse CD27, a 45 kDa transmembrane protein expressed on T cells, B cells, and NK cells. This agonistic antibody stimulates CD27, providing co-stimulation essential for T cell proliferation and immune responses.
- Isotype: Rat IgG2a, κ
- Conjugation: Unconjugated
- Immunogen: Mouse CD27-human IgG1 Fc fusion protein
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
- Applications: In vivo/in vitro CD27 stimulation, immunoprecipitation, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.