Learn More
Bio X Cell Monoclonal Antibody, RM27-3E5, Mouse CD27, 100 MG
Supplier: Bio X Cell BE0348R100MG
The RM27-3E5 monoclonal antibody targets mouse CD27, a 45 kDa type I transmembrane protein of the TNF superfamily, expressed on T cells, B cells, and NK cells. Interaction with CD70 provides T-cell co-stimulation for cell proliferation and antigen-specific T-cell maintenance. Agonistic antibodies like RM27-3E5 are explored as experimental cancer treatments.
- Agonistic antibody stimulates CD27 upon binding
- Purified from animal-free facility cell culture supernatant
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Purity ≥95% (determined by SDS-PAGE)
- Endotoxin ≤1EU/mg (determined by LAL assay)
- Sterility 0.2 μm filtration
- Purification method: Protein G
- Molecular weight: 150 kDa
- Applications: in vivo/in vitro CD27 stimulation, immunoprecipitation, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.