Learn More
Bio X Cell anti-Mouse CD27 Antibody, Clone RM27-3E5, 50 MG
Supplier: Bio X Cell BE0348R050MG
The RM27-3E5 monoclonal antibody reacts with mouse CD27, a 45 kDa type I transmembrane protein and a member of the TNF superfamily. This agonistic antibody stimulates CD27 upon binding, providing co-stimulation in T cells and inducing signaling events essential for immune responses.
- Isotype: rat IgG2a, κ
- Target: mouse CD27
- Host species: rat
- Clone: RM27-3E5
- Purification: protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Storage: store at 4°C, do not freeze
- Applications: in vivo and in vitro CD27 stimulation, immunoprecipitation, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.