Learn More
Bio X Cell anti-mouse CD80, Clone RM80, IgG2a, 25 MG
Supplier: Bio X Cell BE0365R025MG
This monoclonal antibody reacts with mouse CD80 (B7-1), a 60 kDa Ig superfamily member expressed by activated B cells, monocytes, and dendritic cells. It provides costimulatory signals for T cell activation via CD28 and inhibits T cells via CTLA-4, blocking CD80 in vivo.
- Target: mouse CD80 (B7-1)
- Host species: rat
- Isotype: rat IgG2a
- Clone: RM80
- Molecular weight: 150 kDa
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg) by LAL assay
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 µm filtration
- Purification: protein G affinity chromatography
- Storage: store at stock concentration at 4°C; do not freeze
- Reported applications: in vivo CD80 blockade, in vitro CD80 blockade, flow cytometry
- Immunogen: mouse BCL1 cells expressing CD80
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.