Learn More
Bio X Cell anti-Mouse CD80 Monoclonal Antibody, 5 MG
Supplier: Bio X Cell BE0365R005MG
The RM80 monoclonal antibody reacts with mouse CD80 (B7-1), a 60 kDa Ig superfamily member. It provides a costimulatory signal for T cell activation by binding to CD28 and inhibits T cells by binding to CTLA-4. This antibody has been shown to block CD80 in vivo.
- Target: Mouse CD80 (B7-1)
- Clone: RM80
- Isotype: Rat IgG2a
- Host species: Rat
- Reactivity: Mouse
- Immunogen: Mouse BCL1 cells expressing CD80
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg
- Molecular Weight: 150 kDa
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein G affinity chromatography
- Applications: In vivo CD80 blockade, in vitro CD80 blockade, flow cytometry
- Sterility: 0.2 μm filtration
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.