Learn More
Bio X Cell Antibody, Clone RMT3-23, Mouse TIM-3, 1 MG
Supplier: Bio X Cell BE0115R001MG
The InVivoMAb anti-mouse TIM-3 (CD366) antibody reacts with mouse TIM-3 (T cell immunoglobulin and mucin domain-3), also known as CD366. This antibody acts as a TIM-3 receptor antagonist and has been shown to suppress tumor cell growth in a murine sarcoma model.
- Isotype: Rat IgG2a, κ
- Immunogen: Recombinant mouse TIM-3
- Reported applications: in vivo TIM-3 neutralization, in vitro TIM-3 blocking, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
- Conjugation: Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.