Learn More
Bio X Cell SARS-CoV-2 RBD, SARS2-01, Mouse IgG1, 1 MG
Supplier: Bio X Cell BE0357R001MG
The SARS2-01 monoclonal antibody reacts with the receptor binding domain (RBD) in the spike (S) protein of SARS-CoV-2. It has been shown to block the binding of the SARS-CoV-2 S protein to ACE2 in vitro.
- Immunogen: SARS-CoV-2 S and RBD proteins
- Reported applications: In vitro blocking of SARS-CoV-2 S protein, flow cytometry, ELISA
- Purification: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at stock concentration at 4°C, do not freeze
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.