Learn More
Bio X Cell Monoclonal Antibody, SARS-CoV-2 S, SARS2-01, 100 MG
Supplier: Bio X Cell BE0357R100MG
This monoclonal antibody, clone SARS2-01, is a mouse IgG1, κ developed to target the receptor binding domain (RBD) within the SARS-CoV-2 spike (S) protein. It effectively blocks S protein binding to ACE2 in in vitro assays, making it a valuable tool for COVID-19 research.
- Clone: SARS2-01
- Isotype: Mouse IgG1, κ
- Target: SARS-CoV-2 S protein (RBD epitope A)
- Purification: Protein G
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Reported applications: in vitro blocking of SARS-CoV-2 S protein, flow cytometry, ELISA
- Storage: Store at the stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.