Learn More
Bio X Cell Antibody, SARS-CoV-2 S Protein, Mouse IgG1, 50 MG, 50 MG
Supplier: Bio X Cell BE0357R050MG
The SARS2-01 monoclonal antibody targets the receptor binding domain (RBD) in the spike (S) protein of SARS-CoV-2, the virus responsible for COVID-19. The S protein is the primary surface antigen of SARS-CoV-2, with its RBD facilitating host cell entry by binding to angiotensin-converting enzyme 2 (ACE2). This antibody has demonstrated its ability to block the binding of the SARS-CoV-2 S protein to ACE2 in vitro.
- Clone: SARS2-01
- Isotype: Mouse IgG1,
- Immunogen: SARS-CoV-2 S and RBD proteins
- Applications: in vitro blocking of SARS-CoV-2 S protein, Flow cytometry, ELISA
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: 1 EU/mg (0.001 EU/g)
- Purity: 95%
- Sterility: 0.2 m filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.