Learn More
Bio X Cell InVivoMAb anti-SARS-CoV-2 S, Clone SARS2-29, 1 MG
Supplier: Bio X Cell BE0358R001MG
This SARS2-29 monoclonal antibody reacts with the N-terminal domain (NTD) of the SARS-CoV-2 spike (S) protein, which is the main surface antigen mediating host cell entry. It is suitable for applications requiring specific detection of SARS-CoV-2 S protein NTD.
- Reacts with N-terminal domain (NTD) of SARS-CoV-2 S protein
- Clone: SARS2-29
- Isotype: Mouse IgG1, κ
- Applications: Flow cytometry, ELISA
- Immunogen: SARS-CoV-2 S and RBD proteins
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.