Learn More
Bio X Cell Antibody, anti-SARS-CoV-2 NTD, Clone SARS2-29, 100 MG
Supplier: Bio X Cell BE0358R100MG
The SARS2-29 monoclonal antibody reacts with the N-terminal domain (NTD) in the spike (S) protein of SARS-CoV-2. This antibody is designed for research use, targeting the main surface antigen of the virus which mediates host cell entry.
- Reacts with virus
- Isotype mouse IgG1, κ
- Immunogen SARS-CoV-2 S and RBD proteins
- Validated for flow cytometry and ELISA
- Formulation PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin ≤1EU/mg, determined by LAL assay
- Purity ≥95%, determined by SDS-PAGE
- Sterility 0.2 μm filtration
- Molecular weight 150 kDa
- Storage at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.