Learn More
Bio X Cell anti-SARS-CoV-2, Clone SARS2-29, Mouse IgG1, 25 MG
Supplier: Bio X Cell BE0358R025MG
The SARS2-29 monoclonal antibody reacts with the N-terminal domain (NTD) in the spike (S) protein of SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2). SARS-CoV-2 is a positive-sense, single-stranded RNA virus which causes the acute respiratory disease COVID-19. The S protein is the main surface antigen of SARS-CoV-2. The RBD in the S protein specifically binds to angiotensin-converting enzyme 2 (ACE2) on target cells, mediating SARS-CoV-2 host cell entry.
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular Weight: 150 kDa
- Purification: Protein G
- Formulation: PBS, pH 7.0. Contains no stabilizers or preservatives.
- Storage: Antibody solution should be stored at the stock concentration at 4°C. Do not freeze.
- Applications: Flow cytometry, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.