Learn More
Bio X Cell Anti-SARS-CoV-2 S Protein, Mouse IgG1, 50 MG
Supplier: Bio X Cell BE0358R050MG
The SARS2-29 monoclonal antibody reacts with the N-terminal domain (NTD) in the spike (S) protein of SARS-CoV-2, which causes COVID-19. The S protein is the main surface antigen of SARS-CoV-2 and its RBD binds to ACE2 on target cells, mediating host cell entry.
- Isotype: Mouse IgG1, κ
- Immunogen: SARS-CoV-2 S and RBD proteins
- Reported applications: Flow cytometry, ELISA
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1 EU/mg, determined by LAL assay
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein G, from cell culture supernatant in an animal-free facility
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.