Learn More
Bio X Cell Antibody, SARS-CoV-2 RBD, Mouse IgG1, 5 MG
Supplier: Bio X Cell BE0359R005MG
The SARS2-34 monoclonal antibody reacts with the receptor binding domain (RBD) in the spike (S) protein of SARS-CoV-2. It blocks SARS-CoV-2 S protein binding to ACE2 in vitro, mediating host cell entry.
- Isotype: Mouse IgG1, κ
- Isotype control: Mouse IgG1, unknown specificity
- Dilution buffer: pH 7.0
- Unconjugated; conjugation available
- Immunogen: SARS-CoV-2 S and RBD proteins
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Applications: in vitro blocking of SARS-CoV-2 S protein, flow cytometry, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.