Learn More
Bio X Cell Antibody, SARS-CoV-2 S RBD, SARS2-34, 50 MG
Supplier: Bio X Cell BE0359R050MG
The SARS2-34 monoclonal antibody targets the receptor binding domain (RBD) in the spike (S) protein of SARS-CoV-2, blocking its binding to ACE2 in vitro.
- Isotype: Mouse IgG1, κ
- Conjugation: Unconjugated
- Immunogen: SARS-CoV-2 S and RBD proteins
- Reported applications: in vitro blocking of SARS-CoV-2 S protein, Flow cytometry, ELISA
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg) by LAL assay
- Purity: ≥95% by SDS-PAGE
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.