Learn More
Waters Corp Waters Standards, mAb Size Variant, NIST Traceable, SEC Optimized
Catalog No. 502039053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The mAb Size Variant Standard assesses LC systems and columns for separating monoclonal antibody aggregates, monomers, and fragments via SEC. It includes NIST humanized mAb (Reference Material 8671) and IdeS digested fragments F(ab″)2 (~100 kDa) and (Fc/2)2 (~50 kDa). Enables accurate mAb analysis.
  • NIST humanized mAb with IdeS digested fragments F(ab″)2 (~100 kDa) and (Fc/2)2 (~50 kDa).
  • Optimized for SEC to separate mAb aggregates, monomer, fragments.
  • Specs: SEC, optical detection, solid, -20°C ± 10°C (Freezer) storage, 36 months unreconstituted shelf life (1 week reconstituted), NIST traceability.
  • Verifies LC system performance for mAb aggregate detection.
Catalog No. 50-203-9053 Supplier Waters Corp Supplier No. 186009429
The mAb Size Variant Standard assesses LC systems and columns for separating monoclonal antibody aggregates, monomers, and fragments via SEC. It includes NIST humanized mAb (Reference Material 8671) and IdeS digested fragments F(ab″)2 (~100 kDa) and (Fc/2)2 (~50 kDa). Enables accurate mAb analysis.
  • NIST humanized mAb with IdeS digested fragments F(ab″)2 (~100 kDa) and (Fc/2)2 (~50 kDa).
  • Optimized for SEC to separate mAb aggregates, monomer, fragments.
  • Specs: SEC, optical detection, solid, -20°C ± 10°C (Freezer) storage, 36 months unreconstituted shelf life (1 week reconstituted), NIST traceability.
  • Verifies LC system performance for mAb aggregate detection.