Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters mAb Subunit Standard, Reduced, IdeS-Digested, 25 μg

Catalog No. 501528589 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The mAb Subunit Standard is designed for benchmarking Liquid Chromatography (LC) and LC-Mass Spectrometry (LC-MS) techniques. It is also suitable for proficiency testing across various instruments and laboratories, and for system suitability applications. This standard is a filtered and stabilized formulation of reduced, IdeS-digested NIST Reference Material 8671 (NIST mAb), which is a humanized IgG1κ expressed from a murine cell line. Each vial contains approximately 25 μg of the standard.

  • Formulation: Solid
  • Storage condition: –20°C (Freezer)
  • Applications: Environment, protein
  • Units per package: 1 pk
  • Quantity per vial: Approximately 25 μg
  • Source material: Reduced, IdeS-digested NIST Reference Material 8671 (NIST mAb), a humanized IgG1κ expressed from a murine cell line.

Catalog No. 50-152-8589 Supplier Waters Corp Supplier No. 186008927
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The mAb Subunit Standard is designed for benchmarking Liquid Chromatography (LC) and LC-Mass Spectrometry (LC-MS) techniques. It is also suitable for proficiency testing across various instruments and laboratories, and for system suitability applications. This standard is a filtered and stabilized formulation of reduced, IdeS-digested NIST Reference Material 8671 (NIST mAb), which is a humanized IgG1κ expressed from a murine cell line. Each vial contains approximately 25 μg of the standard.

  • Formulation: Solid
  • Storage condition: –20°C (Freezer)
  • Applications: Environment, protein
  • Units per package: 1 pk
  • Quantity per vial: Approximately 25 μg
  • Source material: Reduced, IdeS-digested NIST Reference Material 8671 (NIST mAb), a humanized IgG1κ expressed from a murine cell line.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.