Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Isotype Control, Rat IgG1, Anti-Trinitrophenol, 1 MG

Catalog No. 501528470
Encompass_Preferred

The TNP6A7 monoclonal antibody reacts with trinitrophenol (TNP). Because TNP is not expressed by mammals, this antibody serves as an ideal isotype-matched control for rat IgG1 antibodies in most in vivo and in vitro applications.

  • Clone: TNP6A7
  • Reactivities: Rat
  • Isotype: Rat IgG1, λ
  • Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
  • Conjugation: Unconjugated
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C, do not freeze
  • Package size: 1 mg

Catalog No. 50-152-8470 Supplier Bio X Cell Supplier No. BE0290R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TNP6A7 monoclonal antibody reacts with trinitrophenol (TNP). Because TNP is not expressed by mammals, this antibody serves as an ideal isotype-matched control for rat IgG1 antibodies in most in vivo and in vitro applications.

  • Clone: TNP6A7
  • Reactivities: Rat
  • Isotype: Rat IgG1, λ
  • Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
  • Conjugation: Unconjugated
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C, do not freeze
  • Package size: 1 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.