Learn More
Meridian Life Science MAb to SARS-CoV-2 Nucleocapsid Protein | 1mg
SDP
Supplier:  Meridian Life Science 95471
Encompass
Format: Aff.Pur. | Host/Source: Mouse | Isotype: IgG1 | Clone: B3449M | Tested Apps: EIA, LF, WB
Catalog No. NC1838601