Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-mouse TNFR2 (CD120b), Clone: TR75-54.7, 1 MG

Catalog No. 501528436
Encompass_Preferred

The TR75-54.7 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor Type II (TNFR2), also known as CD120b. It binds to TNFα or LTα ligands, influencing immunoregulation, cell proliferation, differentiation, and apoptosis. The antibody has been reported to block ligand-induced receptor signaling.

  • Isotype: Armenian hamster IgG
  • Immunogen: Recombinant mouse TNFR2
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Applications: In vivo TNFR2 blockade, in vitro TNFR2 blockade

Catalog No. 50-152-8436 Supplier Bio X Cell Supplier No. BE0247R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TR75-54.7 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor Type II (TNFR2), also known as CD120b. It binds to TNFα or LTα ligands, influencing immunoregulation, cell proliferation, differentiation, and apoptosis. The antibody has been reported to block ligand-induced receptor signaling.

  • Isotype: Armenian hamster IgG
  • Immunogen: Recombinant mouse TNFR2
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Applications: In vivo TNFR2 blockade, in vitro TNFR2 blockade

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.