Learn More
Bio X Cell Antibody, Anti-Mouse/Human IL-5, TRFK5 Clone, 1 MG
Supplier: Bio X Cell BE0198R001MG
The TRFK5 monoclonal antibody reacts with mouse and human interleukin-5 (IL-5), a 26 kDa homodimeric cytokine. It neutralizes the bioactivity of natural or recombinant IL-5, stimulating B lymphocyte differentiation and proliferation and increasing IgM and IgA secretion.
- Clone: TRFK5
- Reactivities: Mouse, Human
- Isotype: Rat IgG1, κ
- Reported applications: in vivo IL-5 neutralization, in vivo eosinophil depletion
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.