Learn More
Bio X Cell anti-mouse/human IL-5 Antibody, Rat IgG1, 100 MG
Supplier: Bio X Cell BE0198R100MG
The TRFK5 monoclonal antibody targets mouse and human interleukin-5 (IL-5), a 26 kDa homodimeric cytokine that influences B lymphocyte differentiation and proliferation, and eosinophil activation. It neutralizes the bioactivity of natural or recombinant IL-5.
- Neutralizes natural or recombinant IL-5 bioactivity
- Unconjugated
- Applications: in vivo IL-5 neutralization and eosinophil depletion
- Host species: Rat
- Target species: Mouse, human
- Clone name: TRFK5
- Isotype: IgG1, κ
- Purification: Protein G
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.