Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Tryptic Digestion Standard, Lyophilized, Reduced and Alkylated, NIST mAb, 40 μg

Catalog No. NC1645948 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The mAb Tryptic Digestion Standard supports peptide analysis of proteins by LC-MS, a key method for characterizing biopharmaceuticals like monoclonal antibodies (mAbs). It enables peptide mass information acquisition, sequence confirmation, and identification of post-translational modifications. This standard is ideal for benchmarking LC-MS techniques, proficiency testing, and system suitability in peptide level protein analysis.

  • LC-MS compatible; separation mode: Reversed Phase.
  • Formulation: Solid.
  • Storage: -20°C to +10°C (Freezer); shipping: Ambient; shelf life: 36 months.
  • Type: Digestion.
  • Application: Peptide.
  • Product type: Standards.
  • Units per package: 1 pk.
  • Quantity per vial: 40 μg.
  • Composition: Stabilized, lyophilized, reduced, and alkylated tryptic digest of NIST mAb.
  • Storage after reconstitution: 4°C to 8°C for 7 days.

Catalog No. NC1645948 Supplier Waters Corp Supplier No. 186009126
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The mAb Tryptic Digestion Standard supports peptide analysis of proteins by LC-MS, a key method for characterizing biopharmaceuticals like monoclonal antibodies (mAbs). It enables peptide mass information acquisition, sequence confirmation, and identification of post-translational modifications. This standard is ideal for benchmarking LC-MS techniques, proficiency testing, and system suitability in peptide level protein analysis.

  • LC-MS compatible; separation mode: Reversed Phase.
  • Formulation: Solid.
  • Storage: -20°C to +10°C (Freezer); shipping: Ambient; shelf life: 36 months.
  • Type: Digestion.
  • Application: Peptide.
  • Product type: Standards.
  • Units per package: 1 pk.
  • Quantity per vial: 40 μg.
  • Composition: Stabilized, lyophilized, reduced, and alkylated tryptic digest of NIST mAb.
  • Storage after reconstitution: 4°C to 8°C for 7 days.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.