Learn More
Bio X Cell InVivoMAb anti-PrP Antibody, Clone TW1, 25 MG
Supplier: Bio X Cell BE0350R025MG
The TW1 monoclonal antibody reacts with mouse and human prion protein (PrP), involved in prion neurodegenerative disorders. It binds to both PrPc and PrPSc, blocking PrP-tau interaction and ameliorating tau-related pathology in an Alzheimer's mouse model.
- Isotype: Mouse IgG2a
- Conjugation: Unconjugated
- Immunogen: Non-denatured PK-resistant PrPSc fragment from infected mouse brains
- Reported applications: In vivo PrP blocking, Western blot
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.