Learn More
Bio X Cell Antibody, Mouse CD73, Clone TY/23, 1 MG
Supplier: Bio X Cell BE0209R001MG
The TY/23 monoclonal antibody targets mouse CD73, a 69 kDa GPI-anchored cell-surface protein that converts AMP to adenosine, influencing immune responses. This antibody inhibits the enzymatic activity of CD73.
- Purified by protein G from animal-free cell culture supernatant
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1 EU/mg; purity ≥95%
- Sterility 0.2 μm filtered
- Reported applications include in vivo and in vitro CD73 blockade
- Molecular weight: 150 kDa
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.