Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell anti-LCMV nucleoprotein, VL-4, Rat IgG2a, 1 MG

Catalog No. 501528323
Encompass_Preferred

The VL-4 antibody reacts with lymphocytic choriomeningitis virus (LCMV) nucleoprotein (NP), a 63 kDa structural protein. This antibody stains LCMV-infected cells internally without surface staining and does not react with vaccinia, vesicular stomatitis, or influenza virus-infected cells.

  • Clone: VL-4
  • Isotype: Rat IgG2a, κ
  • Immunogen: LCMV strain WE
  • Molecular weight: 150 kDa
  • Purity: ≥95% by SDS-PAGE
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Applications: Immunofluorescence, flow cytometry
  • Storage: Store at stock concentration at 4°C, do not freeze

Catalog No. 50-152-8323 Supplier Bio X Cell Supplier No. BE0106R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The VL-4 antibody reacts with lymphocytic choriomeningitis virus (LCMV) nucleoprotein (NP), a 63 kDa structural protein. This antibody stains LCMV-infected cells internally without surface staining and does not react with vaccinia, vesicular stomatitis, or influenza virus-infected cells.

  • Clone: VL-4
  • Isotype: Rat IgG2a, κ
  • Immunogen: LCMV strain WE
  • Molecular weight: 150 kDa
  • Purity: ≥95% by SDS-PAGE
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Applications: Immunofluorescence, flow cytometry
  • Storage: Store at stock concentration at 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.