Learn More
Bio X Cell Monoclonal Antibody, anti-mouse IFNg, XMG1.2, 1 MG
Supplier: Bio X Cell BE0055R001MG
The XMG1.2 monoclonal antibody reacts with mouse IFNγ (interferon gamma), a 20 kDa soluble pleiotropic cytokine. It mediates immunoregulatory, anti-proliferative, anti-viral, and proinflammatory activities, and is a neutralizing antibody.
- Clone: XMG1.2
- Target: mouse IFNγ
- Host: Rat; Isotype: Rat IgG1, κ
- Reactivities: Mouse
- Applications: in vivo/in vitro IFNγ neutralization, ELISPOT, Flow cytometry, Western blot
- Immunogen: Recombinant mouse IFNγ
- Formulation: PBS + 0.01% Tween, pH 8.0
- Endotoxin: ≤1EU/mg; Purity: ≥95%
- Sterility: 0.2 μm filtration; Production: Animal-free facility
- Purification: Protein G; Molecular Weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.