Learn More
Bio X Cell Antibody, Anti-mouse TNF alpha XT3.11, 1 MG
Supplier: Bio X Cell BE0058R001MG
The XT3.11 monoclonal antibody targets mouse TNFα, a multifunctional proinflammatory cytokine involved in immunoregulation, cell proliferation, and inflammation. Dysregulation of TNFα is linked to autoimmune diseases, insulin resistance, and cancer.
- Clone: XT3.11
- Reactivities: Mouse
- Isotype: Rat IgG1
- Immunogen: Recombinant mouse TNFα
- Reported applications: in vivo TNFα neutralization, in vitro TNFα neutralization, Western blot
- Formulation: PBS, pH 8.0
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Production: Purified in animal-free facility
- Purification: Protein G
- Storage: Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.