Learn More
Aviva Systems Biology Corp MAB21L1 Antibody - N-terminal region (ARP54782_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54782_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAB21L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAB21L1. Alternate names: CAGR1, FLJ10197, Nbla00126ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53743