Learn More
ATCC MAB35
Catalog No. 502382195
Encompass
mAb35 is a b lymphocyte cell isolated from a mouse. The cell can be used in immunology. The antibody binds to the major immunogenic region of the alpha subunits and cross-reacts well with acetylcholine receptors of a variety of species. Tested and found negative for ectromelia virus (mousepox).
Catalog No. 50-238-2195 Supplier ATCC Supplier No. TIB175
Add to Cart
Add to Cart
mAb35 is a b lymphocyte cell isolated from a mouse. The cell can be used in immunology. The antibody binds to the major immunogenic region of the alpha subunits and cross-reacts well with acetylcholine receptors of a variety of species. Tested and found negative for ectromelia virus (mousepox).