Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MabCaptureC™ MiniChrom™ Pre-packed Column

Catalog No. 5943662005 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
59-436-62005 5 mL
59-436-62001 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-436-62005 Supplier Thermo Scientific™ Supplier No. 5943662005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Packed with MabCaptureC™ Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
  • Direct connection to standard chromatography systems

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.

The pre-packed MabCaptureC™ Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

Features of MabCaptureC™ Affinity Matrix include:

  • High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main resin characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Operating pressure: ≤ 3 bar
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Max. Pressure ≤3 bar
Pore Size Not Applicable
Stationary Phase Agarose bead
Length (Metric) 10 cm
Quantity 1 Column
Particle Size 70 μm
For Use With (Equipment) Chromatography Systems
Type Protein A Column
Diameter (Metric) Inner 0.8 cm
Content And Storage Store at 2°C to 8°C.
Binding Capacity >55 g IgG/L Resin (5 min. residence time and 10% breakthrough), >75 g IgG/L Resin (10 min. residence time and 10% breakthrough)
Product Line MabCaptureC
Ligand Type Engineered rProtein A
Matrix Agarose
Shipping Condition Wet Ice
Target IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain
Column Format Pre-packed
Bed Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.