Learn More
Sigma Aldrich Fine Chemicals Biosciences MabSelectTM Cytiva 17 5199
Catalog No. 502908247
Encompass
MabSelect(TM) is a protein A affinity medium designed for capturing MAbs from large sample volumes.
Catalog No. 50-290-8247 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17519901
MabSelect(TM) is a protein A affinity medium designed for capturing MAbs from large sample volumes.