Learn More
Sigma Aldrich Fine Chemicals Biosciences MAbtrapR Kit Cytiva 17 112
Catalog No. 502904614
Encompass
MAbTrap(R) is an affinity chromatography kit with everything required for small-scale IgG purification from ascites fluid serum and cell culture supernatants.
Catalog No. 50-290-4614 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17112801
MAbTrap(R) is an affinity chromatography kit with everything required for small-scale IgG purification from ascites fluid serum and cell culture supernatants.