Learn More
Electron Microscopy Sciences MAC TEM 40 Thin Film Standards
SDP
Catalog No. 5029299
Encompass_Preferred
These standards are supplied as fine powders that are dispersed onto holey-carbon films and are selected from the range of certified natural and synthetic compounds already in use by us to prepare bulk analysis standards. The holey-carbon films are supported by a 3.05 mm copper grid (400 mesh).
Catalog No. 50-292-99 Supplier Electron Microscopy Sciences Supplier No. 8008040
These standards are supplied as fine powders that are dispersed onto holey-carbon films and are selected from the range of certified natural and synthetic compounds already in use by us to prepare bulk analysis standards. The holey-carbon films are supported by a 3.05 mm copper grid (400 mesh).