Learn More
Selleck Chemical LLC MAC-545496
SDP
Catalog No. 5000002854
Encompass_Preferred
MAC-545496 is a nanomolar glycopeptide-resistance-associated protein R(GraR) inhibitor with strong binding affinity to the full-length GraR protein (Kd 0 1 nM) MAC-545496 can reverse -lactam resistance in methicillin-resistant strains and synergize with CAMPs MAC-545496 shows remarkable activity in macrophages and attenuates S aureus virulence in a G mellonella larvae infection model
Catalog No. 50-000-02854 Supplier Selleck Chemical LLC Supplier No. S67715MG
MAC-545496 is a nanomolar glycopeptide-resistance-associated protein R(GraR) inhibitor with strong binding affinity to the full-length GraR protein (Kd 0 1 nM) MAC-545496 can reverse -lactam resistance in methicillin-resistant strains and synergize with CAMPs MAC-545496 shows remarkable activity in macrophages and attenuates S aureus virulence in a G mellonella larvae infection model